The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 76 - 77 | 12511 | 97 |
XM_006711956.2 | XP_006712019.1 | 76 - 77 | 12616 | 97 | |
XM_011532649.2 | XP_011530951.1 | 76 - 77 | 12465 | 97 | |
XM_011532650.2 | XP_011530952.1 | 76 - 77 | 12466 | 97 | |
XM_011532652.1 | XP_011530954.1 | 75 - 76 | 12385 | 97 | |
XM_011532653.1 | XP_011530955.1 | 75 - 76 | 12364 | 97 | |
XM_011532654.1 | XP_011530956.1 | 75 - 76 | 12244 | 97 | |
XM_011532655.2 | XP_011530957.1 | 72 - 73 | 11533 | 97 | |
XM_011532657.1 | XP_011530959.1 | 70 - 71 | 11272 | 97 | |
XM_017003521.1 | XP_016859010.1 | 74 - 75 | 12238 | 97 | |
XM_017003522.1 | XP_016859011.1 | 71 - 72 | 12337 | 97 | |
XM_017003523.1 | XP_016859012.1 | 71 - 72 | 11504 | 97 | |
GenBank mRNA | AB051484.2 | - | 40 - 41 | 6446 | 97 |
AB290165.1 | - | 42 - 43 | 6650 | 97 | |
AK027182.1 | - | 12 - 13 | 1660 | 97 | |
BC117259.1 | - | 29 - 30 | 4643 | 97 |