Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPO7
Entrez Gene ID
10527
Gene Name
importin 7
Gene Aliases
Imp7, RANBP7
Chromosome Location
Chr.11: 9384622 - 9448127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006391.2 NP_006382.1 21 - 22 2629 70
GenBank mRNA AF098799.1 - 21 - 22 2594 70
AK027487.1 - 1 - 2 133 70
AK027892.1 - 1 - 2 113 70
AK074908.1 - 14 - 15 1660 70
AK312885.1 - 21 - 22 2600 70
BC114929.1 - 21 - 22 2555 70

View Full Product Details