Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198399.1 NP_938409.1 1 - 2 239 77
GenBank mRNA BC017805.1 - 1 - 2 239 77
BG535216.1 - 1 - 2 240 77

View Full Product Details