Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARPP21
- Entrez Gene ID
-
10777
- Gene Name
- cAMP regulated phosphoprotein 21
- Gene Aliases
-
ARPP-21, R3HDM3, RCS, TARPP
- Chromosome Location
-
Chr.3: 35638981 - 35798933 on Build GRCh38
View Full Product Details