The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322253.1 | NP_001309182.1 | 19 - 20 | 3230 | 65 |
NM_001322256.1 | NP_001309185.1 | 17 - 18 | 3120 | 65 | |
NM_001322257.1 | NP_001309186.1 | 17 - 18 | 3022 | 65 | |
NM_015106.3 | NP_055921.2 | 18 - 19 | 3182 | 65 | |
XM_006713062.3 | XP_006713125.1 | 17 - 18 | 2929 | 65 | |
XM_017005987.1 | XP_016861476.1 | 17 - 18 | 2958 | 65 | |
XM_017005988.1 | XP_016861477.1 | 18 - 19 | 4518 | 65 | |
GenBank mRNA | AB018352.2 | - | 15 - 16 | 2587 | 65 |
AK023824.1 | - | 12 - 13 | 3382 | 65 | |
AK025049.1 | - | 7 - 8 | 1028 | 65 | |
AK304881.1 | - | 13 - 14 | 2318 | 65 | |
AK308456.1 | - | 16 - 17 | 2719 | 65 | |
BC001474.2 | - | 11 - 12 | 1910 | 65 | |
BC024298.1 | - | 15 - 16 | 2546 | 65 |