Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNA1
Entrez Gene ID
1942
Gene Name
ephrin A1
Gene Aliases
B61, ECKLG, EFL1, EPLG1, LERK-1, LERK1, TNFAIP4
Chromosome Location
Chr.1: 155127873 - 155134910 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182685.1 NP_872626.1 2 - 3 491 80
GenBank mRNA AK289589.1 - 2 - 3 488 80
AL833641.1 - 2 - 3 489 80

View Full Product Details