Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA2D2
Entrez Gene ID
9254
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Gene Aliases
CACNA2D
Chromosome Location
Chr.3: 50362613 - 50504244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005505.2 NP_001005505.1 11 - 12 1325 58
NM_001174051.2 NP_001167522.1 11 - 12 1325 58
NM_001291101.1 NP_001278030.1 11 - 12 1121 58
NM_006030.3 NP_006021.2 11 - 12 1325 58
XM_005265581.4 XP_005265638.1 11 - 12 1199 58
XM_011534243.2 XP_011532545.1 11 - 12 1199 58
GenBank mRNA AB011130.1 - 11 - 12 1152 58
AF040709.1 - 11 - 12 1331 58
AF042792.1 - 12 - 13 1312 58
AF042793.1 - 11 - 12 1128 58
AJ251367.1 - 11 - 12 1151 58
AJ251368.1 - 11 - 12 1151 58
BC152438.1 - 11 - 12 1152 58

View Full Product Details