Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A42
Entrez Gene ID
284439
Gene Name
solute carrier family 25 member 42
Gene Aliases
-
Chromosome Location
Chr.19: 19063961 - 19113030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321544.1 NP_001308473.1 3 - 4 304 62
NM_178526.4 NP_848621.2 3 - 4 340 62
XM_005259861.3 XP_005259918.1 4 - 5 434 62
XM_011527937.2 XP_011526239.1 3 - 4 316 62
XM_017026670.1 XP_016882159.1 2 - 3 577 62
GenBank mRNA AK303785.1 - 2 - 3 366 62
BC045598.1 - 3 - 4 337 62
FN356975.1 - 3 - 4 315 62

View Full Product Details