The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004386.2 | NP_004377.2 | 9 - 10 | 3230 | 98 |
GenBank mRNA | AB210004.1 | - | 9 - 10 | 3174 | 98 |
AF026547.1 | - | 8 - 9 | 3132 | 98 | |
AK297395.1 | - | 3 - 4 | 1536 | 98 | |
AK309138.1 | - | 3 - 4 | 1702 | 98 |