The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005261203.4 | XP_005261260.1 | 1 - 2 | 581 | 77 |
GenBank mRNA | AL079407.1 | - | 1 - 2 | 370 | 77 |