Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EBAG9
Entrez Gene ID
9166
Gene Name
estrogen receptor binding site associated, antigen, 9
Gene Aliases
EB9, PDAF
Chromosome Location
Chr.8: 109539700 - 109565996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278938.1 NP_001265867.1 5 - 6 538 106
NM_004215.4 NP_004206.1 5 - 6 825 106
NM_198120.2 NP_936056.1 5 - 6 658 106
XM_017013960.1 XP_016869449.1 5 - 6 1021 106
GenBank mRNA AB007619.1 - 5 - 6 706 106
AF006265.1 - 5 - 6 656 106
AK290651.1 - 5 - 6 647 106
AL515533.3 - 5 - 6 716 106
AY515724.1 - 5 - 6 616 106
AY653072.1 - 4 - 5 423 106
BC005249.1 - 5 - 6 736 106
BC017729.1 - 5 - 6 609 106
BC022506.1 - 5 - 6 723 106
BG616704.1 - 5 - 6 539 106
CR456984.1 - 4 - 5 423 106

View Full Product Details