Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF14
Entrez Gene ID
9678
Gene Name
PHD finger protein 14
Gene Aliases
-
Chromosome Location
Chr.7: 10973834 - 11188565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033436.1 15 - 16 2313 70
XM_017012813.1 XP_016868302.1 16 - 17 3138 70
XM_017012814.1 XP_016868303.1 16 - 17 3138 70
GenBank mRNA AA699404.1 - 2 - 3 70
AA837024.1 - 2 - 3 70
AK295461.1 - 15 - 16 2296 70

View Full Product Details