The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005261430.2 | XP_005261487.1 | 10 - 11 | 1558 | 68 |
XM_006724187.2 | XP_006724250.1 | 10 - 11 | 1570 | 68 |