Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFOX2
Entrez Gene ID
23543
Gene Name
RNA binding protein, fox-1 homolog 2
Gene Aliases
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location
Chr.22: 35738736 - 36028892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031695.2 NP_001026865.1 9 - 10 1267 62
NM_001082576.1 NP_001076045.1 9 - 10 1255 62
NM_001082578.1 NP_001076047.1 10 - 11 1227 62
NM_001082579.1 NP_001076048.1 10 - 11 1224 62
XM_005261428.2 XP_005261485.1 10 - 11 1570 62
XM_005261429.2 XP_005261486.1 10 - 11 1567 62
XM_005261431.2 XP_005261488.1 10 - 11 1570 62
XM_005261432.2 XP_005261489.1 10 - 11 1567 62
XM_005261435.1 XP_005261492.1 9 - 10 1271 62
XM_005261437.1 XP_005261494.1 9 - 10 1274 62
XM_006724185.2 XP_006724248.1 10 - 11 1582 62
XM_006724186.2 XP_006724249.1 10 - 11 1579 62
XM_006724189.3 XP_006724252.1 10 - 11 1275 62
XM_006724190.1 XP_006724253.1 10 - 11 969 62
XM_006724191.1 XP_006724254.1 9 - 10 1274 62
XM_006724192.2 XP_006724255.1 9 - 10 1104 62
XM_006724193.3 XP_006724256.1 10 - 11 1582 62
XM_006724194.2 XP_006724257.1 9 - 10 1046 62
XM_017028686.1 XP_016884175.1 10 - 11 961 62
XM_017028687.1 XP_016884176.1 10 - 11 1142 62
XM_017028689.1 XP_016884178.1 10 - 11 1035 62
XM_017028690.1 XP_016884179.1 9 - 10 1024 62
XM_017028691.1 XP_016884180.1 9 - 10 1021 62
XM_017028692.1 XP_016884181.1 10 - 11 1304 62
XM_017028693.1 XP_016884182.1 9 - 10 1262 62
XM_017028694.1 XP_016884183.1 9 - 10 1259 62
XM_017028696.1 XP_016884185.1 9 - 10 1262 62
XM_017028697.1 XP_016884186.1 10 - 11 2051 62
GenBank mRNA AB649123.1 - 9 - 10 902 62
AF229058.1 - 9 - 10 1072 62
AK055213.1 - 10 - 11 1188 62
AK291460.1 - 9 - 10 1151 62
AK314632.1 - 9 - 10 1086 62
AM419009.1 - 9 - 10 914 62
AY072786.1 - 10 - 11 1037 62
BC013115.1 - 8 - 9 1025 62
BC025281.1 - 9 - 10 1090 62
CR456559.1 - 9 - 10 956 62
DQ778625.1 - 10 - 11 1115 62

View Full Product Details