Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSBP
Entrez Gene ID
5007
Gene Name
oxysterol binding protein
Gene Aliases
OSBP1
Chromosome Location
Chr.11: 59574398 - 59616144 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002556.2 NP_002547.1 12 - 13 2544 70
GenBank mRNA AF185696.1 - 12 - 13 2544 70
AK299071.1 - 8 - 9 988 70
AK310538.1 - 6 - 7 802 70
BC011581.1 - 12 - 13 2132 70
BC063121.1 - 6 - 7 903 70
M86917.1 - 12 - 13 2544 70

View Full Product Details