The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252148.1 | NP_001239077.1 | 2 - 3 | 909 | 102 |
NM_001252150.1 | NP_001239079.1 | 2 - 3 | 909 | 102 | |
NM_001252151.1 | NP_001239080.1 | 2 - 3 | 336 | 102 | |
NM_018375.4 | NP_060845.2 | 2 - 3 | 909 | 102 | |
GenBank mRNA | AK002136.1 | - | 2 - 3 | 885 | 102 |
AK222547.1 | - | 2 - 3 | 887 | 102 | |
AK293222.1 | - | 2 - 3 | 329 | 102 | |
AY358687.1 | - | 2 - 3 | 355 | 102 | |
AY780789.1 | - | 2 - 3 | 207 | 102 | |
BC047682.1 | - | 2 - 3 | 839 | 102 | |
BC064383.1 | - | 2 - 3 | 624 | 102 | |
BP269907.1 | - | 1 - 2 | 111 | 102 | |
FJ423549.1 | - | 2 - 3 | 207 | 102 |