The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252148.1 | NP_001239077.1 | 4 - 5 | 1195 | 84 |
NM_001252150.1 | NP_001239079.1 | 5 - 6 | 1264 | 84 | |
NM_001252151.1 | NP_001239080.1 | 5 - 6 | 691 | 84 | |
NM_001252152.1 | NP_001239081.1 | 6 - 7 | 679 | 84 | |
NM_018375.4 | NP_060845.2 | 5 - 6 | 1264 | 84 | |
XM_005267834.1 | XP_005267891.1 | 4 - 5 | 1066 | 84 | |
XM_017021426.1 | XP_016876915.1 | 6 - 7 | 1405 | 84 | |
GenBank mRNA | AK002136.1 | - | 5 - 6 | 1240 | 84 |
AK222547.1 | - | 5 - 6 | 1242 | 84 | |
AK293222.1 | - | 5 - 6 | 684 | 84 | |
AK316381.1 | - | 6 - 7 | 668 | 84 | |
AY358687.1 | - | 5 - 6 | 710 | 84 | |
AY780789.1 | - | 4 - 5 | 493 | 84 | |
BC047682.1 | - | 5 - 6 | 1194 | 84 | |
BC064383.1 | - | 5 - 6 | 979 | 84 | |
BP269907.1 | - | 4 - 5 | 466 | 84 | |
FJ423549.1 | - | 5 - 6 | 562 | 84 |