Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN3B
Entrez Gene ID
55800
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
ATFB16, BRGDA7, HSA243396, SCNB3
Chromosome Location
Chr.11: 123629187 - 123654607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040151.1 NP_001035241.1 3 - 4 625 75
NM_018400.3 NP_060870.1 2 - 3 1024 75
XM_011542897.1 XP_011541199.1 3 - 4 625 75
GenBank mRNA AB032984.1 - 3 - 4 265 75
AJ243396.2 - 2 - 3 593 75
AK314513.1 - 3 - 4 612 75
AL136589.1 - 2 - 3 1021 75
BC117282.1 - 2 - 3 311 75
BC126265.1 - 2 - 3 311 75
BP202832.1 - 2 - 3 499 75

View Full Product Details