The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018644.3 | NP_061114.2 | 2 - 3 | 225 | 86 |
XM_017017550.1 | XP_016873039.1 | 1 - 2 | 113 | 86 | |
GenBank mRNA | AB029396.1 | - | 2 - 3 | 139 | 86 |