The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282614.1 | NP_001269543.1 | 9 - 10 | 1324 | 133 |
GenBank mRNA | AF440758.1 | - | 9 - 10 | 1269 | 133 |
BX374313.2 | - | 1 - 2 | 133 |