The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018365.2 | NP_060835.1 | 2 - 3 | 394 | 157 |
GenBank mRNA | AK002084.1 | - | 2 - 3 | 394 | 157 |
AK057542.1 | - | 1 - 2 | 208 | 157 | |
BC031046.1 | - | 2 - 3 | 307 | 157 | |
BC034991.2 | - | 2 - 3 | 361 | 157 |