The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018365.2 | NP_060835.1 | 6 - 7 | 1066 | 67 |
GenBank mRNA | AK002084.1 | - | 6 - 7 | 1066 | 67 |
BC031046.1 | - | 6 - 7 | 979 | 67 | |
BC034991.2 | - | 6 - 7 | 1033 | 67 |