The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018365.2 | NP_060835.1 | 7 - 8 | 1170 | 143 |
GenBank mRNA | AK002084.1 | - | 7 - 8 | 1170 | 143 |
AK057542.1 | - | 6 - 7 | 970 | 143 | |
BC031046.1 | - | 7 - 8 | 1083 | 143 | |
BC034991.2 | - | 7 - 8 | 1137 | 143 |