Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC49
Entrez Gene ID
54839
Gene Name
leucine rich repeat containing 49
Gene Aliases
PGs4
Chromosome Location
Chr.15: 70852671 - 71050097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199017.2 NP_001185946.1 13 - 14 2061 100
NM_001199018.2 NP_001185947.1 14 - 15 2135 100
NM_001284357.1 NP_001271286.1 14 - 15 2093 100
NM_017691.4 NP_060161.2 13 - 14 2046 100
XM_005254492.4 XP_005254549.1 12 - 13 1762 100
XM_011521715.2 XP_011520017.1 15 - 16 2396 100
XM_011521717.2 XP_011520019.1 14 - 15 1977 100
XM_011521718.2 XP_011520020.1 13 - 14 1874 100
XM_017022357.1 XP_016877846.1 14 - 15 2676 100
XM_017022358.1 XP_016877847.1 13 - 14 1806 100
XM_017022359.1 XP_016877848.1 13 - 14 1851 100
GenBank mRNA AK000163.1 - 13 - 14 1838 100
AK097752.1 - 14 - 15 1812 100
AK123672.1 - 14 - 15 2050 100
AK125732.1 - 15 - 16 1940 100
AK225137.1 - 13 - 14 1851 100
AK295547.1 - 13 - 14 1672 100
AK301971.1 - 14 - 15 1743 100
BC037982.1 - 13 - 14 1657 100

View Full Product Details