The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199017.2 | NP_001185946.1 | 5 - 6 | 967 | 95 |
NM_001199018.2 | NP_001185947.1 | 6 - 7 | 1041 | 95 | |
NM_001284357.1 | NP_001271286.1 | 6 - 7 | 999 | 95 | |
NM_017691.4 | NP_060161.2 | 5 - 6 | 952 | 95 | |
XM_005254492.4 | XP_005254549.1 | 4 - 5 | 668 | 95 | |
XM_011521715.2 | XP_011520017.1 | 7 - 8 | 1302 | 95 | |
XM_011521717.2 | XP_011520019.1 | 6 - 7 | 883 | 95 | |
XM_011521718.2 | XP_011520020.1 | 5 - 6 | 780 | 95 | |
XM_017022357.1 | XP_016877846.1 | 6 - 7 | 1582 | 95 | |
XM_017022358.1 | XP_016877847.1 | 5 - 6 | 712 | 95 | |
XM_017022359.1 | XP_016877848.1 | 5 - 6 | 757 | 95 | |
GenBank mRNA | AK000163.1 | - | 5 - 6 | 747 | 95 |
AK097752.1 | - | 6 - 7 | 721 | 95 | |
AK123672.1 | - | 6 - 7 | 959 | 95 | |
AK225137.1 | - | 5 - 6 | 760 | 95 | |
AK295547.1 | - | 5 - 6 | 578 | 95 | |
AK301971.1 | - | 6 - 7 | 652 | 95 | |
BC037982.1 | - | 5 - 6 | 566 | 95 |