Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1A
Entrez Gene ID
4068
Gene Name
SH2 domain containing 1A
Gene Aliases
DSHP, EBVS, IMD5, LYP, MTCP1, SAP, SAP/SH2D1A, XLP, XLPD, XLPD1
Chromosome Location
Chr.X: 124346282 - 124373160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002351.4 NP_002342.1 3 - 4 705 113
GenBank mRNA AB586694.1 - 3 - 4 446 113
AF072930.1 - 3 - 4 430 113
AF073019.1 - 3 - 4 430 113
AF100539.1 - 3 - 4 343 113
AF100540.1 - 2 - 3 334 113
AF100542.1 - 3 - 4 321 113
AF100543.1 - 2 - 3 279 113
AK311645.1 - 2 - 3 204 113
AK311911.1 - 3 - 4 379 113
AL023657.1 - 3 - 4 643 113
BC020732.1 - 3 - 4 437 113
CR542031.1 - 3 - 4 344 113
CR542043.1 - 3 - 4 344 113

View Full Product Details