Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFX
Entrez Gene ID
7543
Gene Name
zinc finger protein, X-linked
Gene Aliases
ZNF926
Chromosome Location
Chr.X: 24148994 - 24216255 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178085.1 NP_001171556.1 2 - 3 171 89
NM_003410.3 NP_003401.2 2 - 3 163 89
XM_006724513.3 XP_006724576.1 2 - 3 441 89
XM_006724515.3 XP_006724578.1 2 - 3 443 89
XM_011545578.1 XP_011543880.1 2 - 3 323 89
XM_011545579.1 XP_011543881.1 2 - 3 228 89
XM_017029788.1 XP_016885277.1 1 - 2 236 89
XM_017029794.1 XP_016885283.1 2 - 3 269 89
GenBank mRNA BC136308.1 - 1 - 2 34 89
BC136309.1 - 1 - 2 34 89
DA848249.1 - 2 - 3 163 89
M30608.1 - 1 - 2 114 89

View Full Product Details