The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320923.1 | NP_001307852.1 | 3 - 4 | 212 | 77 |
NM_001321852.1 | NP_001308781.1 | 2 - 3 | 215 | 77 | |
NM_001321853.1 | NP_001308782.1 | 4 - 5 | 474 | 77 | |
NM_001321854.1 | NP_001308783.1 | 3 - 4 | 390 | 77 | |
NM_001321855.1 | NP_001308784.1 | 3 - 4 | 393 | 77 | |
NM_001321856.1 | NP_001308785.1 | 2 - 3 | 128 | 77 | |
NM_001321857.1 | NP_001308786.1 | 2 - 3 | 309 | 77 | |
NM_002227.3 | NP_002218.2 | 2 - 3 | 309 | 77 | |
XM_017001212.1 | XP_016856701.1 | 2 - 3 | 156 | 77 | |
GenBank mRNA | AB209057.1 | - | 1 - 2 | 65 | 77 |
AK314257.1 | - | 3 - 4 | 214 | 77 | |
BC009079.1 | - | 2 - 3 | 244 | 77 | |
BC062431.1 | - | 2 - 3 | 244 | 77 | |
BC111401.1 | - | 2 - 3 | 274 | 77 | |
M64174.1 | - | 1 - 2 | 34 | 77 |