The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130158.1 | NP_001123630.1 | 21 - 22 | 2548 | 72 |
NM_001161819.1 | NP_001155291.1 | 21 - 22 | 2386 | 72 | |
NM_012223.3 | NP_036355.2 | 21 - 22 | 2548 | 72 | |
XM_005246572.1 | XP_005246629.1 | 21 - 22 | 2412 | 72 | |
XM_017004156.1 | XP_016859645.1 | 21 - 22 | 2521 | 72 | |
XM_017004157.1 | XP_016859646.1 | 21 - 22 | 2521 | 72 | |
XM_017004158.1 | XP_016859647.1 | 19 - 20 | 2137 | 72 | |
GenBank mRNA | AB290176.1 | - | 21 - 22 | 2565 | 72 |
AJ001381.1 | - | 4 - 5 | 366 | 72 | |
AJ001382.1 | - | 3 - 4 | 326 | 72 | |
AK000160.1 | - | 17 - 18 | 1882 | 72 | |
AK074978.1 | - | 3 - 4 | 288 | 72 | |
BC053558.1 | - | 21 - 22 | 2386 | 72 |