Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135248.1 NP_001128720.1 1 - 2 90 75
NM_002403.3 NP_002394.1 1 - 2 90 75
GenBank mRNA AK222751.1 - 1 - 2 52 75
BC015039.1 - 1 - 2 90 75
DN996554.1 - 1 - 2 40 75

View Full Product Details