The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199163.1 | NP_001186092.1 | 8 - 9 | 1081 | 99 |
NM_002805.5 | NP_002796.4 | 8 - 9 | 946 | 99 | |
GenBank mRNA | AF035309.1 | - | - | 1367 | 99 |
AK095836.1 | - | 7 - 8 | 2180 | 99 | |
AK098484.1 | - | - | 2731 | 99 | |
AK290758.1 | - | 8 - 9 | 1044 | 99 | |
AK291878.1 | - | 8 - 9 | 918 | 99 | |
BC001932.1 | - | 8 - 9 | 897 | 99 | |
BC002367.2 | - | 8 - 9 | 876 | 99 | |
D44467.1 | - | 8 - 9 | 876 | 99 |