The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180991.4 | NP_851322.3 | 1 - 2 | 640 | 94 |
GenBank mRNA | AF401643.1 | - | 1 - 2 | 353 | 94 |
AK128854.1 | - | 1 - 2 | 368 | 94 | |
AY273896.2 | - | 1 - 2 | 640 | 94 | |
BX647349.1 | - | 1 - 2 | 533 | 94 |