The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180991.4 | NP_851322.3 | 2 - 3 | 906 | 111 |
XM_011543372.1 | XP_011541674.1 | 4 - 5 | 557 | 111 | |
GenBank mRNA | AF401643.1 | - | 2 - 3 | 619 | 111 |
AK128854.1 | - | 2 - 3 | 634 | 111 | |
AY273896.2 | - | 2 - 3 | 906 | 111 | |
BX647349.1 | - | 2 - 3 | 799 | 111 |