The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002738.6 | NP_002729.2 | 14 - 15 | 1814 | 97 |
NM_212535.2 | NP_997700.1 | 14 - 15 | 1814 | 97 | |
GenBank mRNA | BC036472.1 | - | 14 - 15 | 1774 | 97 |
M13975.1 | - | 14 - 15 | 1681 | 97 | |
X06318.1 | - | 14 - 15 | 1753 | 97 | |
X07109.1 | - | 14 - 15 | 1753 | 97 |