The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012231.4 | NP_036363.2 | 1 - 2 | 789 | 130 |
NM_015866.4 | NP_056950.2 | 1 - 2 | 789 | 130 | |
GenBank mRNA | U17838.1 | - | 1 - 2 | 788 | 130 |