The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073135.1 | 1 - 2 | 96 | 117 | |
XM_006724354.3 | XP_006724417.1 | 1 - 2 | 83 | 117 | |
XM_011530486.2 | XP_011528788.1 | 1 - 2 | 83 | 117 | |
XM_017029042.1 | XP_016884531.1 | 1 - 2 | 83 | 117 | |
XM_017029049.1 | XP_016884538.1 | 1 - 2 | 83 | 117 | |
XM_017029051.1 | XP_016884540.1 | 1 - 2 | 83 | 117 | |
XM_017029058.1 | XP_016884547.1 | 1 - 2 | 61 | 117 | |
XM_017029059.1 | XP_016884548.1 | 1 - 2 | 87 | 117 | |
XM_017029063.1 | XP_016884552.1 | 1 - 2 | 87 | 117 | |
XM_017029064.1 | XP_016884553.1 | 1 - 2 | 61 | 117 | |
GenBank mRNA | BC028625.1 | - | 1 - 2 | 92 | 117 |
BC050285.1 | - | 1 - 2 | 69 | 117 |