Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBN3
Entrez Gene ID
84467
Gene Name
fibrillin 3
Gene Aliases
-
Chromosome Location
Chr.19: 8065402 - 8149846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321431.1 NP_001308360.1 2 - 3 305 128
NM_032447.4 NP_115823.3 2 - 3 580 128
XM_017027372.1 XP_016882861.1 2 - 3 304 128
XM_017027373.1 XP_016882862.1 2 - 3 563 128
XM_017027374.1 XP_016882863.1 2 - 3 563 128
XM_017027375.1 XP_016882864.1 2 - 3 563 128
XM_017027376.1 XP_016882865.1 2 - 3 563 128
XM_017027379.1 XP_016882868.1 2 - 3 563 128
GenBank mRNA AB053450.2 - 1 - 2 186 128
AY165863.1 - 2 - 3 580 128
AY165864.1 - 2 - 3 305 128
AY165865.1 - 1 - 2 451 128
BC153882.1 - 1 - 2 186 128

View Full Product Details