The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013437.1 | NP_001013455.1 | 7 - 8 | 1052 | 91 |
NM_031216.3 | NP_112493.2 | 7 - 8 | 1052 | 91 | |
XM_005258152.4 | XP_005258209.1 | 6 - 7 | 895 | 91 | |
XM_011525742.2 | XP_011524044.1 | 8 - 9 | 1108 | 91 | |
XM_011525743.2 | XP_011524045.1 | 7 - 8 | 955 | 91 | |
XM_011525744.1 | XP_011524046.1 | 8 - 9 | 1108 | 91 | |
XM_011525745.2 | XP_011524047.1 | 7 - 8 | 876 | 91 | |
XM_011525746.2 | XP_011524048.1 | 7 - 8 | 1158 | 91 | |
XM_017026025.1 | XP_016881514.1 | 6 - 7 | 901 | 91 | |
XM_017026026.1 | XP_016881515.1 | 6 - 7 | 972 | 91 | |
XM_017026027.1 | XP_016881516.1 | 7 - 8 | 1222 | 91 | |
GenBank mRNA | AF136976.1 | - | 7 - 8 | 1020 | 91 |
AF255625.1 | - | 7 - 8 | 956 | 91 | |
AF431970.1 | - | 7 - 8 | 956 | 91 | |
AF514996.1 | - | 7 - 8 | 916 | 91 | |
AK056940.1 | - | 7 - 8 | 1015 | 91 | |
AK226075.1 | - | 4 - 5 | 3442 | 91 | |
AK291226.1 | - | 7 - 8 | 1015 | 91 | |
AK314403.1 | - | 7 - 8 | 1026 | 91 | |
BC012430.1 | - | 7 - 8 | 1006 | 91 | |
BX538037.1 | - | 2 - 3 | 2761 | 91 |