The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013437.1 | NP_001013455.1 | 8 - 9 | 1208 | 138 |
XM_005258152.4 | XP_005258209.1 | 7 - 8 | 1051 | 138 | |
XM_011525742.2 | XP_011524044.1 | 9 - 10 | 1264 | 138 | |
XM_011525743.2 | XP_011524045.1 | 8 - 9 | 1111 | 138 | |
XM_011525745.2 | XP_011524047.1 | 8 - 9 | 1032 | 138 | |
XM_011525746.2 | XP_011524048.1 | 8 - 9 | 1314 | 138 | |
XM_017026026.1 | XP_016881515.1 | 7 - 8 | 1128 | 138 | |
GenBank mRNA | AF431970.1 | - | 8 - 9 | 1112 | 138 |
AK056940.1 | - | 8 - 9 | 1171 | 138 |