The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319084.1 | NP_001306013.1 | 5 - 6 | 773 | 92 |
NM_001319086.1 | NP_001306015.1 | 5 - 6 | 1181 | 92 | |
NM_003707.2 | NP_003698.1 | 5 - 6 | 714 | 92 | |
XM_005247841.3 | XP_005247898.1 | 5 - 6 | 635 | 92 | |
XM_011513248.1 | XP_011511550.1 | 5 - 6 | 761 | 92 | |
XM_011513249.2 | XP_011511551.1 | 5 - 6 | 768 | 92 | |
XM_017007356.1 | XP_016862845.1 | 5 - 6 | 768 | 92 | |
XM_017007357.1 | XP_016862846.1 | 5 - 6 | 638 | 92 | |
GenBank mRNA | AB012122.1 | - | 5 - 6 | 663 | 92 |
AB451224.1 | - | 5 - 6 | 610 | 92 | |
AB451347.1 | - | 5 - 6 | 610 | 92 | |
AF070735.1 | - | 5 - 6 | 686 | 92 | |
AF099084.1 | - | 5 - 6 | 674 | 92 | |
AJ010058.1 | - | 5 - 6 | 686 | 92 | |
AK222563.1 | - | 5 - 6 | 695 | 92 | |
AK222575.1 | - | 5 - 6 | 686 | 92 | |
AK312290.1 | - | 5 - 6 | 710 | 92 | |
BC002993.1 | - | 5 - 6 | 681 | 92 | |
BC012886.1 | - | 5 - 6 | 643 | 92 | |
BT007057.1 | - | 5 - 6 | 610 | 92 | |
DQ469310.1 | - | 5 - 6 | 610 | 92 | |
Y18418.1 | - | 5 - 6 | 706 | 92 |