The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042432.1 | NP_001035897.1 | 1 - 2 | 276 | 93 |
NM_001286104.1 | NP_001273033.1 | 1 - 2 | 276 | 93 | |
GenBank mRNA | AK090709.1 | - | 2 - 3 | 1271 | 93 |
AK294250.1 | - | 1 - 2 | 46 | 93 | |
AK302138.1 | - | 1 - 2 | 44 | 93 | |
AK313002.1 | - | 1 - 2 | 48 | 93 | |
BP252176.1 | - | 1 - 2 | 56 | 93 | |
DA703317.1 | - | 1 - 2 | 276 | 93 | |
DB101039.1 | - | 1 - 2 | 276 | 93 |