The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015891.2 | NP_056975.1 | 1 - 2 | 212 | 90 |
GenBank mRNA | AB208903.1 | - | 1 - 2 | 186 | 90 |
AF015044.1 | - | 1 - 2 | 179 | 90 | |
AF038392.1 | - | 1 - 2 | 212 | 90 | |
AF061241.1 | - | 1 - 2 | 185 | 90 | |
AK222721.1 | - | 1 - 2 | 209 | 90 | |
AK295055.1 | - | 1 - 2 | 205 | 90 | |
AK314601.1 | - | 1 - 2 | 208 | 90 | |
BC111061.1 | - | 2 - 3 | 289 | 90 | |
BC117145.1 | - | 1 - 2 | 209 | 90 | |
BC126114.1 | - | 1 - 2 | 209 | 90 |