The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002581.4 | NP_002572.2 | 13 - 14 | 3989 | 78 |
XM_006717129.2 | XP_006717192.1 | 11 - 12 | 2063 | 78 | |
XM_017014784.1 | XP_016870273.1 | 12 - 13 | 3915 | 78 | |
GenBank mRNA | AK300129.1 | - | 7 - 8 | 920 | 78 |
BC078657.1 | - | 13 - 14 | 3619 | 78 | |
X68280.1 | - | 13 - 14 | 3380 | 78 |