The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002581.4 | NP_002572.2 | 8 - 9 | 3245 | 83 |
XM_006717129.2 | XP_006717192.1 | 6 - 7 | 1319 | 83 | |
XM_017014784.1 | XP_016870273.1 | 8 - 9 | 3285 | 83 | |
GenBank mRNA | AK300128.1 | - | 6 - 7 | 1472 | 83 |
AK300129.1 | - | 2 - 3 | 176 | 83 | |
BC078657.1 | - | 8 - 9 | 2875 | 83 | |
BU158714.1 | - | 2 - 3 | 364 | 83 | |
X68280.1 | - | 8 - 9 | 2636 | 83 |