The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144875.1 | NP_001138347.1 | 4 - 5 | 674 | 69 |
NM_001144876.1 | NP_001138348.1 | 3 - 4 | 450 | 69 | |
NM_001308235.1 | NP_001295164.1 | 4 - 5 | 866 | 69 | |
NM_001308236.1 | NP_001295165.1 | 4 - 5 | 689 | 69 | |
NM_024872.2 | NP_079148.2 | 4 - 5 | 645 | 69 | |
XM_005265985.3 | XP_005266042.1 | 4 - 5 | 702 | 69 | |
XM_005265987.1 | XP_005266044.1 | 4 - 5 | 702 | 69 | |
XM_011534654.2 | XP_011532956.1 | 4 - 5 | 579 | 69 | |
XM_011534655.2 | XP_011532957.1 | 4 - 5 | 755 | 69 | |
XM_011534656.2 | XP_011532958.1 | 4 - 5 | 556 | 69 | |
XM_011534657.2 | XP_011532959.1 | 3 - 4 | 406 | 69 | |
XM_017009883.1 | XP_016865372.1 | 2 - 3 | 377 | 69 | |
XM_017009884.1 | XP_016865373.1 | 2 - 3 | 377 | 69 | |
GenBank mRNA | AK026223.1 | - | 4 - 5 | 640 | 69 |
AK097258.1 | - | 3 - 4 | 406 | 69 | |
BC004564.2 | - | 4 - 5 | 633 | 69 | |
BC004867.2 | - | 2 - 3 | 741 | 69 |