Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TLE3
- Entrez Gene ID
-
7090
- Gene Name
- transducin like enhancer of split 3
- Gene Aliases
-
ESG, ESG3, GRG3, HsT18976
- Chromosome Location
-
Chr.15: 70047790 - 70097935 on Build GRCh38
View Full Product Details