The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195193.1 | NP_001182122.1 | 11 - 12 | 2120 | 85 |
NM_002482.3 | NP_002473.2 | 13 - 14 | 2312 | 85 | |
NM_152298.3 | NP_689511.2 | 12 - 13 | 1295 | 85 | |
XM_005270888.2 | XP_005270945.1 | 14 - 15 | 2332 | 85 | |
XM_005270889.2 | XP_005270946.1 | 13 - 14 | 1315 | 85 | |
GenBank mRNA | AK092829.1 | - | 7 - 8 | 3333 | 85 |
AK131249.1 | - | 8 - 9 | 2427 | 85 | |
AK222778.1 | - | 13 - 14 | 2255 | 85 | |
AK222816.1 | - | 13 - 14 | 2255 | 85 | |
AK291637.1 | - | 13 - 14 | 2255 | 85 | |
AK298893.1 | - | 11 - 12 | 2063 | 85 | |
AK299584.1 | - | 12 - 13 | 2119 | 85 | |
BC003113.1 | - | 12 - 13 | 1213 | 85 | |
BC010105.2 | - | 13 - 14 | 2240 | 85 | |
BC011580.1 | - | 13 - 14 | 2238 | 85 | |
BC053608.1 | - | 12 - 13 | 1218 | 85 | |
BT006757.1 | - | 12 - 13 | 1136 | 85 | |
M97856.1 | - | 13 - 14 | 2235 | 85 |