The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289050.1 | NP_001275979.1 | 6 - 7 | 1057 | 82 |
NM_016609.4 | NP_057693.3 | 7 - 8 | 1449 | 82 | |
NM_020372.3 | NP_065105.2 | 6 - 7 | 1182 | 82 | |
XM_005267747.4 | XP_005267804.1 | 7 - 8 | 3525 | 82 | |
XM_017021361.1 | XP_016876850.1 | 7 - 8 | 3522 | 82 | |
XM_017021362.1 | XP_016876851.1 | 7 - 8 | 3520 | 82 | |
GenBank mRNA | AJ243653.1 | - | 4 - 5 | 497 | 82 |
BC020565.1 | - | 6 - 7 | 1001 | 82 | |
BC090870.1 | - | 6 - 7 | 733 | 82 | |
BC111015.1 | - | 5 - 6 | 596 | 82 | |
BX161416.1 | - | 7 - 8 | 1408 | 82 | |
DQ658848.1 | - | 6 - 7 | 827 | 82 |