The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278541.1 | NP_001265470.1 | 4 - 5 | 443 | 122 |
NM_001872.4 | NP_001863.3 | 4 - 5 | 443 | 122 | |
XM_017020393.1 | XP_016875882.1 | 4 - 5 | 386 | 122 | |
GenBank mRNA | AB011969.1 | - | 4 - 5 | 395 | 122 |
AK290829.1 | - | 4 - 5 | 391 | 122 | |
BC007057.1 | - | 4 - 5 | 400 | 122 | |
BT006936.1 | - | 4 - 5 | 376 | 122 | |
M75106.1 | - | 4 - 5 | 395 | 122 |