Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
Chromosome Location
Chr.2: 25377220 - 25673647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320933.1 NP_001307862.1 10 - 11 1343 85
NM_001320934.1 NP_001307863.1 10 - 11 1343 85
NM_033148.3 NP_149160.1 10 - 11 1343 85
NM_183361.2 NP_899205.1 10 - 11 1343 85
XM_011532676.1 XP_011530978.1 10 - 11 1260 85
XM_017003529.1 XP_016859018.1 10 - 11 1260 85
XM_017003530.1 XP_016859019.1 10 - 11 1260 85
XM_017003531.1 XP_016859020.1 10 - 11 1260 85
XM_017003532.1 XP_016859021.1 10 - 11 1260 85
XM_017003533.1 XP_016859022.1 10 - 11 1260 85
XM_017003534.1 XP_016859023.1 10 - 11 1260 85
XM_017003535.1 XP_016859024.1 10 - 11 1260 85
XM_017003536.1 XP_016859025.1 10 - 11 1260 85
XM_017003539.1 XP_016859028.1 9 - 10 1117 85
XM_017003540.1 XP_016859029.1 9 - 10 1174 85
XM_017003541.1 XP_016859030.1 9 - 10 1158 85
XM_017003542.1 XP_016859031.1 9 - 10 1112 85
XM_017003543.1 XP_016859032.1 8 - 9 1006 85
XM_017003544.1 XP_016859033.1 9 - 10 1112 85
XM_017003545.1 XP_016859034.1 8 - 9 1006 85
GenBank mRNA BC016655.1 - 10 - 11 1236 85
BT009805.1 - 9 - 10 1080 85
BX389926.1 - 4 - 5 463 85
R15062.1 - 2 - 3 193 85
Y12712.1 - 10 - 11 1263 85

View Full Product Details